ENA405194823 Enamine

7-oxo-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD20647861
  • Cas Number : 34102-76-6
  • Molecular Weight : 180.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135182
Availability details for
Quantity
  • Availability
  • Add to Cart