ENA405194829 Enamine

2,5,5-trimethylhexanoic acid

  • MDL number MFCD14676691
  • Cas Number : 53705-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135199
Availability details for
Quantity
  • Availability
  • Add to Cart