ENA405194830 Enamine

2-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]-3-methylbutanoic acid

  • MDL number MFCD18433227
  • Cas Number : 180181-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135202
Availability details for
Quantity
  • Availability
  • Add to Cart