ENA405194835 Enamine

4-chloro-2-[(4-chlorophenyl)sulfanyl]benzoic acid

  • MDL number MFCD19154977
  • Cas Number : 20092-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135212

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135212
Availability details for
Quantity
  • Availability
  • Add to Cart