ENA405194891 Enamine

1-[(dimethylamino)methyl]cyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD25970505
  • Cas Number : 1461706-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135232
Availability details for
Quantity
  • Availability
  • Add to Cart