ENA405194892 Enamine

2-[5-(methoxymethyl)-1,2,4-oxadiazol-3-yl]azepane hydrochloride

  • MDL number MFCD25970506
  • Cas Number : 1461714-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135234

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135234
Availability details for
Quantity
  • Availability
  • Add to Cart