ENA405194899 Enamine

2-(4-fluorophenyl)-N-methyloxan-4-amine hydrochloride

  • MDL number MFCD25970512
  • Cas Number : 1461714-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135286
Availability details for
Quantity
  • Availability
  • Add to Cart