ENA405194903 Enamine

methyl 1-(azetidin-3-yl)-1H-1,2,3-triazole-4-carboxylate hydrochloride

  • MDL number MFCD25970516
  • Cas Number : 1461715-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135337
Availability details for
Quantity
  • Availability
  • Add to Cart