ENA405194916 Enamine

2-[5-(propan-2-yl)-1,2,4-oxadiazol-3-yl]morpholine hydrochloride

  • MDL number MFCD25970530
  • Cas Number : 1461705-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135484
Availability details for
Quantity
  • Availability
  • Add to Cart