ENA405194921 Enamine

2-{6-bromo-8-methylimidazo[1,2-a]pyridin-2-yl}ethan-1-amine hydrochloride

  • MDL number MFCD25970535
  • Cas Number : 1461706-94-4
  • Molecular Weight : 290.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(6-bromo-8-methylimidazo[1,2-a]pyridin-2-yl)ethan-1-amine hydrochloride
Inchi Code 1S/C10H12BrN3.ClH/c1-7-4-8(11)5-14-6-9(2-3-12)13-10(7)14;/h4-6H,2-3,12H2,1H3;1H
Inchi Key YZEDFEGFPUMWSF-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-135549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135549
Availability details for
Quantity
  • Availability
  • Add to Cart