ENA405194924 Enamine

3-[5-(methoxymethyl)-1,2,4-oxadiazol-3-yl]morpholine hydrochloride

  • MDL number MFCD25970538
  • Cas Number : 1461715-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135589
Availability details for
Quantity
  • Availability
  • Add to Cart