ENA405194926 Enamine

5-carbamoylfuran-3-carboxylic acid

  • MDL number MFCD25970541
  • Cas Number : 1461706-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135680
Availability details for
Quantity
  • Availability
  • Add to Cart