ENA405194935 Enamine

1-amino-3-(2,4-difluorophenyl)propan-2-ol hydrochloride

  • MDL number MFCD25970550
  • Cas Number : 1461705-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135708
Availability details for
Quantity
  • Availability
  • Add to Cart