ENA405194938 Enamine

2-(4-methyl-3-oxopiperazin-1-yl)acetic acid hydrochloride

  • MDL number MFCD25970553
  • Cas Number : 1205125-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135768
Availability details for
Quantity
  • Availability
  • Add to Cart