ENA405194940 Enamine

4-methyl-2-[(4-methylpyrimidin-2-yl)disulfanyl]pyrimidine dihydrochloride

  • MDL number MFCD25970555
  • Cas Number : 1461705-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35231

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35231
Availability details for
Quantity
  • Availability
  • Add to Cart