ENA405194946 Enamine

2-(1,3-thiazol-2-yl)propan-1-amine

  • MDL number MFCD09891725
  • Cas Number : 92932-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135380

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135380
Availability details for
Quantity
  • Availability
  • Add to Cart