ENA405194947 Enamine

7-bromo-1-azatricyclo[7.3.1.0,5,13]trideca-5,7,9(13)-trien-2-one

  • MDL number MFCD19334787
  • Cas Number : 57369-19-4
  • Molecular Weight : 266.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-135381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135381
Availability details for
Quantity
  • Availability
  • Add to Cart