ENA405194949 Enamine

5-(methylsulfanyl)pentan-1-ol

  • MDL number MFCD09926783
  • Cas Number : 57774-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135283
Availability details for
Quantity
  • Availability
  • Add to Cart