ENA405194955 Enamine

2-cyclopropyl-5-hydrazinyl-1,3,4-thiadiazole

  • MDL number MFCD11890858
  • Cas Number : 299168-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-135392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135392
Availability details for
Quantity
  • Availability
  • Add to Cart