ENA405194957 Enamine

4-chloro-6-(4-methoxyphenoxy)-2-methylpyrimidine

  • MDL number MFCD17349044
  • Cas Number : 1275401-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135395
Availability details for
Quantity
  • Availability
  • Add to Cart