ENA405194961 Enamine

1-(5-methyl-1,3,4-thiadiazol-2-yl)ethan-1-amine

  • MDL number MFCD21289345
  • Cas Number : 1368716-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135230

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-135230
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart