ENA405194982 Enamine

2-[1-(methoxymethyl)cyclopropyl]acetonitrile

  • MDL number MFCD21247049
  • Cas Number : 1188283-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135476
Availability details for
Quantity
  • Availability
  • Add to Cart