ENA405194984 Enamine

methyl 2-(cyclobutylamino)benzoate

  • MDL number MFCD16108134
  • Cas Number : 1247084-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135306
Availability details for
Quantity
  • Availability
  • Add to Cart