ENA405194986 Enamine

5-bromo-1,4-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD11869815
  • Cas Number : 1142188-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135481
Availability details for
Quantity
  • Availability
  • Add to Cart