ENA405194990 Enamine

1-(3-bromo-4-methoxyphenyl)-2,2,2-trifluoroethan-1-amine

  • MDL number MFCD07384639
  • Cas Number : 1039867-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135487
Availability details for
Quantity
  • Availability
  • Add to Cart