ENA405194992 Enamine

2-{[(naphthalen-1-yl)methyl]amino}pentanoic acid

  • MDL number MFCD12692168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135489
Availability details for
Quantity
  • Availability
  • Add to Cart