ENA405195000 Enamine

1-(3,4,5-trimethoxyphenyl)piperazine

  • MDL number MFCD09027417
  • Cas Number : 38869-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135532
Availability details for
Quantity
  • Availability
  • Add to Cart