ENA405195024 Enamine

1-(4-chlorophenyl)-2-(pyridin-3-yl)ethan-1-one

  • MDL number MFCD02271259
  • Cas Number : 58158-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135575
Availability details for
Quantity
  • Availability
  • Add to Cart