ENA405195026 Enamine

1-methyl-1,2,3,4-tetrahydroquinolin-4-amine

  • MDL number MFCD13179152
  • Cas Number : 851390-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135577

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135577
Availability details for
Quantity
  • Availability
  • Add to Cart