ENA405195028 Enamine

4-hydroxyquinazoline-7-carbonitrile

  • MDL number MFCD15527383
  • Cas Number : 882976-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135579
Availability details for
Quantity
  • Availability
  • Add to Cart