ENA405195035 Enamine

4-methoxy-2-methylbutan-2-amine

  • MDL number MFCD16127064
  • Cas Number : 889765-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135659
Availability details for
Quantity
  • Availability
  • Add to Cart