ENA405195038 Enamine

4-[({[4-(pyrrolidin-1-ylmethyl)phenyl]methyl}amino)methyl]benzonitrile dihydrochloride

  • MDL number MFCD25970540
  • Cas Number : 1384724-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 267...269
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135668

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135668
Availability details for
Quantity
  • Availability
  • Add to Cart