ENA405195039 Enamine

2-bromo-5-chloro-4-methylpyridine

  • MDL number MFCD11846275
  • Cas Number : 885267-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135673
Availability details for
Quantity
  • Availability
  • Add to Cart