ENA405195040 Enamine

2-[(4-chlorophenyl)sulfanyl]-4-methylbenzoic acid

  • MDL number MFCD17378778
  • Cas Number : 1274871-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215...217
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135674

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135674
Availability details for
Quantity
  • Availability
  • Add to Cart