ENA405195044 Enamine

methyl 5-amino-3-methyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD09953591
  • Cas Number : 23286-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135321
Availability details for
Quantity
  • Availability
  • Add to Cart