ENA405195045 Enamine

2-(cyclopentylsulfanyl)-4-methylbenzoic acid

  • MDL number MFCD18270813
  • Cas Number : 1340160-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135681
Availability details for
Quantity
  • Availability
  • Add to Cart