ENA405195049 Enamine

2-chloro-5-[(4-methyl-1H-pyrazol-1-yl)methyl]pyridine

  • MDL number MFCD12800660
  • Cas Number : 1183842-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135324
Availability details for
Quantity
  • Availability
  • Add to Cart