ENA405195050 Enamine

2-[(3-fluorophenyl)sulfanyl]-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD18970644
  • Cas Number : 53542-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135325

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135325
Availability details for
Quantity
  • Availability
  • Add to Cart