ENA405195052 Enamine

[1-(ethanesulfonyl)piperidin-4-yl]methanamine hydrochloride

  • MDL number MFCD18378190
  • Cas Number : 1286273-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135327

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135327
Availability details for
Quantity
  • Availability
  • Add to Cart