ENA405195054 Enamine

2-bromo-3-(cyclopropylmethoxy)pyridine

  • MDL number MFCD21162928
  • Cas Number : 910606-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135329
Availability details for
Quantity
  • Availability
  • Add to Cart