ENA405195153 Enamine

4,5-dichlorobenzene-1,2-dicarboxylic acid

  • MDL number MFCD00002493
  • Cas Number : 56962-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17989

Safety Information

signalword Warning
safety_statement 22-24/25
msds https://www.enaminestore.com/catalog/EN300-17989
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart