ENA405195306 Enamine

1-(hydrazinecarbonyl)formamide

  • MDL number MFCD00008004
  • Cas Number : 515-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-218
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-20421

Safety Information

signalword Danger
pcodes P210,P240,P241,P264,P270,P280,P301+P310,P330,P370+P378,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-20421
pictograms GHS02,GHS06
hstatements H228,H301
Availability details for
Quantity
  • Availability
  • Add to Cart