ENA405195416 Enamine

2-chloro-N-(5-chloropyridin-2-yl)acetamide

  • MDL number MFCD00297021
  • Cas Number : 90931-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34064

Safety Information

msds https://www.enaminestore.com/catalog/EN300-34064
Availability details for
Quantity
  • Availability
  • Add to Cart