ENA405195421 Enamine

4-(aminomethyl)benzene-1-sulfonamide

  • MDL number MFCD00072084
  • Cas Number : 138-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35248
Availability details for
Quantity
  • Availability
  • Add to Cart