ENA405195440 Enamine

1-chloro-3-[(1E)-2-nitroprop-1-en-1-yl]benzene

  • MDL number MFCD01317576
  • Cas Number : 19394-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55540
Availability details for
Quantity
  • Availability
  • Add to Cart