ENA405195447 Enamine

4-(3-aminophenyl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD09933328
  • Cas Number : 1016771-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64544
Availability details for
Quantity
  • Availability
  • Add to Cart