ENA405195463 Enamine

1-(4-benzyl-1,4-diazepan-1-yl)ethan-1-one

  • MDL number MFCD25970556
  • Cas Number : 192869-86-6
  • Molecular Weight : 232.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74726
Availability details for
Quantity
  • Availability
  • Add to Cart