ENA405195468 Enamine

1-methyl-1H-imidazole-2-sulfonamide

  • MDL number MFCD16687578
  • Cas Number : 89517-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77789
Availability details for
Quantity
  • Availability
  • Add to Cart