ENA405195476 Enamine

3,4-dimethylcyclohexan-1-amine

  • MDL number MFCD19205340
  • Cas Number : 59477-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83961
Availability details for
Quantity
  • Availability
  • Add to Cart