ENA405195477 Enamine

2-amino-2-cyanoacetamide

  • MDL number MFCD00040532
  • Cas Number : 6719-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-124
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-84364

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P311,P363,P403+P233,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-84364
pictograms GHS05,GHS06
hstatements H314,H331
Availability details for
Quantity
  • Availability
  • Add to Cart